Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Formaldehyde Test Strips, MQuant®

Formaldehyde Test Strips, MQuant®

Formaldehyde Test Strips, 1pc, Triazole, 10 - 100 mg/L HCHO, Plastic Box(es), 100
Catalog # CA1.10036.0001
Supplier:   MilliporeSigma
Supplier #:  1.10036.0001
Formaldehyde Test Strips, MQuant®
Catalog # CA1.10036.0001
Supplier:  MilliporeSigma
CAS Number:  
Formaldehyde Test Strips, MQuant®

Formaldehyde Test Strips, 1pc, Triazole, 10 - 100 mg/L HCHO, Plastic Box(es), 100

CAS Number:  
Price
$264.50
Log in for availability
Each Case of 40
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

MQuant™ test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.

  • Easy to handle: Just dip, wait and measure visually against colour scale
  • Safe: No auxiliary reagents necessary
  • Immediate results within 2 minutes enables fast decisions
  • Portable: Can be used inside or outside the laboratory

Formaldehyde reacts with 4-amino-3-hydrazino5-mercapto-1,2,4-triazole to form a purple-red tetrazine. The formaldehyde concentration is measured semiquantitatively by visual comparison of the reaction zone of the test strip with the fields of a color scale.

Gradation: 10 - 20 - 40 - 60 - 100 mg/l HCHO

Applications: Disinfectant and rinsing solutions (e.g. laundries), aqueous solutions.

Documents

Specifications

Specifications

  • Méthode chimique:
    Triazole
  • Type de conditionnement:
    Plastic Box(es)
  • Désignation:
    Formaldehyde Test Strips
  • Taille:
    1pc
  • Range:
    10 - 100 mg/L HCHO
  • No. of Tests:
    100

Specifications

Certains produits peuvent apparaître comme restreints

Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.

Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)

 

Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.