Sodium Hydroxide
Product Details & Documents
CAS Number: 1310-73-2
Formula: NaOH
Density: 2.13 g/mL
Boiling and Freezing Point: 1390°C, 318°C
Solubility: Water and Alcohol
Synonyms: Caustic Soda
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: This product is corrosive and toxic.
Specifications
Specifications
- Désignation:Sodium Hydroxide, Solution
- Taille:500 mL
- Chemical/Physical Specifications:3.0 M
- Grade:Laboratory
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



