Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sodium Fluoride | Avantor

Sodium Fluoride

Sodium Fluoride, Reagent, 100 g
Catalog # 470302-540
Supplier:   Ward's Science
Fournisseur #:  SS0535-100GR
Sodium Fluoride
Catalog # 470302-540
Supplier:  Ward's Science
CAS Number:  
Sodium Fluoride
Sodium Fluoride

Sodium Fluoride, Reagent, 100 g

CAS Number:  
Price
$32.00
Log in pour la disponibilité
Each (100g)
La quantité minimale de commande est de 1
Product Details & Documents

CAS Number: 7681-49-4
Formula Weight: 41.99
Formula: NaF
Hazard Info: Toxic
Density (g/mL): 2.78
Boiling Point (°C): 1704
Freezing Point (°C): 993
Solubility: Water and Alcohol
Synonyms: Fluoridine
Shelf Life (months): 36
Storage: Blue

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Spécifications

Spécifications

  • Désignation:
    Sodium Fluoride
  • Taille:
    100 g
  • Chemical/Physical Specifications:
    Powder
  • Grade:
    Reagent

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.