Sodium Fluoride
Sodium Fluoride, Solution, Laboratory, 50 mL
Supplier: Ward's Science
Fournisseur #: SS0534-50ML
Jump to:
Product Details & Documents
CAS Number: 7681-49-4
Formula Weight: 41.99
Formula: NaF
Hazard Info: Toxic
Density (g/mL): 2.78
Boiling Point (°C): 1704
Freezing Point (°C): 993
Solubility: Water and Alcohol
Synonyms: Fluoridine
Shelf Life (months): 36
Storage: Blue
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Spécifications
Spécifications
- Désignation:Sodium Fluoride, Solution
- Taille:50 mL
- Chemical/Physical Specifications:0.1 M
- Grade:Laboratory



