Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Universal Plug Stoppers, Globe Scientific | Avantor

Universal Plug Stoppers, Globe Scientific

Lavender
Catalog # 10142-456
Supplier:   Globe Scientific
Fournisseur #:  5529L
Universal Plug Stoppers, Globe Scientific
Catalog # 10142-456
Supplier:  Globe Scientific
Universal Plug Stoppers, Globe Scientific
Universal Plug Stoppers, Globe Scientific

Lavender

Price
$94.13
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 1,000
Product Details & Documents

These universal plug stoppers fit most 10 mm, 12 mm, 13 mm, and 16 mm tubes

  • Low density polyethylene (LDPE)
  • Assorted colors available
  • Add the convenience of 1 cap to fit multiple size tubes

These caps are available in 7 different colors.

Ordering information: Stoppers are packaged 1000 per bag.

Spécifications
  • Color:
    Lavender
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.