Universal Plug Stoppers, Globe Scientific
Universal Plug Stoppers, Globe Scientific
Green
Catalog # 10142-454
Supplier:   Globe Scientific
Supplier #:  5529G
Universal Plug Stoppers, Globe Scientific
Catalog # 10142-454
Supplier:  Globe Scientific
Universal Plug Stoppers, Globe Scientific
Universal Plug Stoppers, Globe Scientific
Green
Price
$94.13
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Pack of 1,000
Product Details & Documents
These universal plug stoppers fit most 10 mm, 12 mm, 13 mm, and 16 mm tubes
  • Low density polyethylene (LDPE)
  • Assorted colors available
  • Add the convenience of 1 cap to fit multiple size tubes
These caps are available in 7 different colors.
Ordering information: Stoppers are packaged 1000 per bag.
Spécifications
  • Color:
    Green