Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Potassium Nitrite | Avantor

Potassium Nitrite

Potassium Nitrite, Reagent, 500 g
Catalog # 470302-208
Supplier:   Ward's Science
Fournisseur #:  PP0670-500GR
Potassium Nitrite
Catalog # 470302-208
Supplier:  Ward's Science
CAS Number:  
Potassium Nitrite
Potassium Nitrite

Potassium Nitrite, Reagent, 500 g

CAS Number:  
Price
$157.60
Log in pour la disponibilité
Each (500g)
La quantité minimale de commande est de 1
Product Details & Documents

CAS Number: 7758-09-0
Formula Weight: 85.11
Formula: KNO2
Hazard Info: Oxidizer, Flammable, Irritant
Density (g/mL): 1.915
Boiling Point (°C): Explodes at 538
Solubility: Water
Synonyms: Nitrous Acid, Potassium Salt
Shelf Life (months): 36
Storage: Yellow

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Spécifications

Spécifications

  • Désignation:
    Potassium Nitrite
  • Taille:
    500 g
  • Chemical/Physical Specifications:
    Crystal
  • Grade:
    Reagent

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.