Potassium Nitrite
Potassium Nitrite, Reagent, 100 g
Supplier: Ward's Science
: PP0670-100GR
Jump to:
Product Details & Documents
CAS Number: 7758-09-0
Formula Weight: 85.11
Formula: KNO2
Hazard Info: Oxidizer, Flammable, Irritant
Density (g/mL): 1.915
Boiling Point (°C): Explodes at 538
Solubility: Water
Synonyms: Nitrous Acid, Potassium Salt
Shelf Life (months): 36
Storage: Yellow
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Spécifications
Spécifications
- Désignation:Potassium Nitrite
- Taille:100 g
- Chemical/Physical Specifications:Crystal
- Grade:Reagent



