Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Potassium Nitrate

Potassium Nitrate, Reagent, 100 g
Catalog # 470302-200
Supplier:   Ward's Science
 :  PP0655-100GR
Potassium Nitrate
Catalog # 470302-200
Supplier:  Ward's Science
CAS Number:  
Potassium Nitrate
Potassium Nitrate

Potassium Nitrate, Reagent, 100 g

CAS Number:  
Price
18,25 $
Log in pour la disponibilité
Each (100g)
 
Product Details & Documents

CAS Number: 7757-79-1
Formula: KNO3
Density: 2.1 g/mL
Boiling and Freezing Point: 400°C, 333°C
Solubility: Water, Glycerin, and Alcohol
Synonyms: Potash Nitrate, Saltpeter
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

 : This product is flammable oxidizer and irritant.

Spécifications

Spécifications

  • Désignation:
    Potassium Nitrate
  • Taille:
    100 g
  • Chemical/Physical Specifications:
    Crystal
  • Grade:
    Reagent

Spécifications