Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Potassium Hydroxide | Avantor

Potassium Hydroxide

Potassium Hydroxide, Laboratory, 500 g
Catalog # 470302-134
Supplier:   Ward's Science
Fournisseur #:  PP0590-500G
Potassium Hydroxide
Catalog # 470302-134
Supplier:  Ward's Science
CAS Number:  
Potassium Hydroxide
Potassium Hydroxide

Potassium Hydroxide, Laboratory, 500 g

CAS Number:  
Price
$52.00
Log in pour la disponibilité
Each (500g)
La quantité minimale de commande est de 1
Product Details & Documents

CAS Number: 1310-58-3
Formula Weight: 56.11
Formula: KOH
Hazard Info: Corrosive, Irritant, Toxic
Density (g/mL): 2.044
Boiling Point (°C): 1320
Freezing Point (°C): 361
Solubility: Water, Alcohol, and Glycerin
Synonyms: Caustic Potash, Potassium Hydrate
Shelf Life (months): 36
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Spécifications

Spécifications

  • Désignation:
    Potassium Hydroxide
  • Taille:
    500 g
  • Chemical/Physical Specifications:
    Flake
  • Grade:
    Laboratory

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.