Potassium Hydroxide
Potassium Hydroxide, Laboratory, 2.5 kg
Supplier: Ward's Science
Fournisseur #: PP0590-2.5KG
Jump to:
Product Details & Documents
CAS Number: 1310-58-3
Formula Weight: 56.11
Formula: KOH
Hazard Info: Corrosive, Irritant, Toxic
Density (g/mL): 2.044
Boiling Point (°C): 1320
Freezing Point (°C): 361
Solubility: Water, Alcohol, and Glycerin
Synonyms: Caustic Potash, Potassium Hydrate
Shelf Life (months): 36
Storage: White
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Spécifications
Spécifications
- Désignation:Potassium Hydroxide
- Taille:2.5 kg
- Chemical/Physical Specifications:Flake
- Grade:Laboratory



