Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Potassium Hydroxide | Avantor

Potassium Hydroxide

Potassium Hydroxide, QuickPrep, Laboratory, Makes 500 mL
Catalog # 470302-130
Supplier:   Ward's Science
Fournisseur #:  PP0570-CONC
Potassium Hydroxide
Catalog # 470302-130
Supplier:  Ward's Science
CAS Number:  
Potassium Hydroxide
Potassium Hydroxide

Potassium Hydroxide, QuickPrep, Laboratory, Makes 500 mL

CAS Number:  
Price
$19.50
Log in pour la disponibilité
Each (50ml)
La quantité minimale de commande est de 1
Product Details & Documents

CAS Number: 1310-58-3
Formula Weight: 56.11
Formula: KOH
Hazard Info: Corrosive, Irritant, Toxic
Density (g/mL): 2.044
Boiling Point (°C): 1320
Freezing Point (°C): 361
Solubility: Water, Alcohol, and Glycerin
Synonyms: Caustic Potash, Potassium Hydrate
Shelf Life (months): 36
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Spécifications

Spécifications

  • Désignation:
    Potassium Hydroxide, QuickPrep
  • Taille:
    Makes 500 mL
  • Chemical/Physical Specifications:
    1 M
  • Grade:
    Laboratory

Spécifications

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.