Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Potassium Dichromate | Avantor

Potassium Dichromate

Potassium Dichromate, Laboratory, 500 g
Catalog # 470302-110
Supplier:   Ward's Science
Fournisseur #:  PP0516-500G
Potassium Dichromate
Catalog # 470302-110
Supplier:  Ward's Science
CAS Number:  
Potassium Dichromate
Potassium Dichromate

Potassium Dichromate, Laboratory, 500 g

CAS Number:  
Price
$137.00
Log in pour la disponibilité
Each (500g)
La quantité minimale de commande est de 1
Product Details & Documents

CAS Number: 7778-50-9
Formula Weight: 294.19
Formula: K2Cr2O7
Density (g/mL): 2.67
Freezing Point (°C): 398
Solubility: Water
Synonyms: Potassium Bichromate
Shelf Life (months): 36
Storage: Yellow

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Spécifications

Spécifications

  • Désignation:
    Potassium Dichromate
  • Taille:
    500 g
  • Chemical/Physical Specifications:
    Granular
  • Grade:
    Laboratory

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.