Potassium Dichromate
Potassium Dichromate, Solution, Laboratory, 50 mL
: PP0521-50ML
CAS Number: 7778-50-9
Formula Weight: 294.19
Formula: K2Cr2O7
Density (g/mL): 2.67
Freezing Point (°C): 398
Solubility: Water
Synonyms: Potassium Bichromate
Shelf Life (months): 36
Storage: Yellow
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
- Désignation:Potassium Dichromate, Solution
- Taille:50 mL
- Chemical/Physical Specifications:0.1 M
- Grade:Laboratory
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



