Potassium Chlorate
Potassium Chlorate, Reagent, 500 g
Supplier: Ward's Science
: PP0439-500GR
Jump to:
Product Details & Documents
CAS Number: 3811-04-9
Formula: KClO3
Density: 2.34 g/mL
Boiling and Freezing Point: 400°C, 368°C
Solubility: Boiling Water
Synonyms: Chlorate of Potash, Chloric Acid Potassium Salt, Potassium Oxymuriate
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
: This product is an explosive hazard, oxidizer, and irritant.
Spécifications
Spécifications
- Désignation:Potassium Chlorate
- Taille:500 g
- Chemical/Physical Specifications:Crystal
- Grade:Reagent



