Potassium Chlorate
CAS Number: 3811-04-9
Formula: KClO3
Density: 2.34 g/mL
Boiling and Freezing Point: 400°C, 368°C
Solubility: Boiling Water
Synonyms: Chlorate of Potash, Chloric Acid Potassium Salt, Potassium Oxymuriate
Shelf Life: 36 Months
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
: This product is an explosive hazard, oxidizer, and irritant.
- Désignation:Potassium Chlorate
- Taille:2.5 kg
- Chemical/Physical Specifications:Crystal
- Grade:Laboratory
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



