Strontium Nitrate
Product Details & Documents
CAS Number: 10042-76-9
Formula Weight: 211.63
Formula: Sr(NO3)2
Hazard Info: Oxidizer, Flammable, Toxic, Irritant
Density (g/mL): 2.986
Freezing Point (°C): 570
Solubility: Water and Alcohol
Shelf Life (months): 36
Storage: Yellow
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Spécifications
Spécifications
- Désignation:Strontium Nitrate
- Taille:100 g
- Chemical/Physical Specifications:Crystal
- Grade:Reagent
Spécifications
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



