Strontium Nitrate
Strontium Nitrate, Laboratory, 4 x 25 g
Supplier #: SS1010-PK/4
CAS Number: 10042-76-9
Formula Weight: 211.63
Formula: Sr(NO3)2
Hazard Info: Oxidizer, Flammable, Toxic, Irritant
Density (g/mL): 2.986
Freezing Point (°C): 570
Solubility: Water and Alcohol
Shelf Life (months): 36
Storage: Yellow
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
- Désignation:Strontium Nitrate
- Taille:4 x 25 g
- Chemical/Physical Specifications:Granular
- Grade:Laboratory



