Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Strontium Nitrate

Strontium Nitrate, Laboratory, 100 g
Catalog # 470302-796
Supplier:   Ward's Science
 :  SS1010-100G
Strontium Nitrate
Catalog # 470302-796
Supplier:  Ward's Science
CAS Number:  
Strontium Nitrate
Strontium Nitrate

Strontium Nitrate, Laboratory, 100 g

CAS Number:  
Price
$23.05
Log in pour la disponibilité
Each (100g)
 
Product Details & Documents

CAS Number: 10042-76-9
Formula Weight: 211.63
Formula: Sr(NO3)2
Hazard Info: Oxidizer, Flammable, Toxic, Irritant
Density (g/mL): 2.986
Freezing Point (°C): 570
Solubility: Water and Alcohol
Shelf Life (months): 36
Storage: Yellow

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

 : This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Spécifications

Spécifications

  • Désignation:
    Strontium Nitrate
  • Taille:
    100 g
  • Chemical/Physical Specifications:
    Granular
  • Grade:
    Laboratory

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.