Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sodium Silicate

Sodium Silicate

Sodium Silicate, Laboratory, 500 g
Catalog # 470302-682
Supplier:   Ward's Science
Supplier #:  SS0805-500G
Sodium Silicate
Catalog # 470302-682
Supplier:  Ward's Science
CAS Number:  
Sodium Silicate
Sodium Silicate

Sodium Silicate, Laboratory, 500 g

CAS Number:  
Price
48,45 $
Log in pour la disponibilité
Each (500g)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 10213-79-3
Formula Weight: 212.03
Formula: Na2SiO3·5H2O
Density (g/mL): 1.81
Freezing Point (°C): 72.2
Synonyms: Disodium Trioxosilicate Pentahydrate
Shelf Life (months): 36
Storage: White

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Spécifications

Spécifications

  • Désignation:
    Sodium Silicate
  • Taille:
    500 g
  • Chemical/Physical Specifications:
    Crystal
  • Grade:
    Laboratory

Spécifications