Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Treston Lab Carts | Avantor

Treston Lab Carts

2 shelf lab cart, 330 lbs.
Catalog # 10129-036
Supplier:   Treston
Fournisseur #:  TRTA4102
Treston Lab Carts
Catalog # 10129-036
Supplier:  Treston
Treston Lab Carts
Treston Lab Carts

2 shelf lab cart, 330 lbs.

Price
$1,564.48
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents

TRTA lab carts are ideal for storing medical devices, instruments and supplies, and moving them to where they are needed.

  • Versatile and multi-purpose
  • Physically ergonomic for effortless cart movement

The height adjustment range of the shelves is 7.87 to 27.56".

Ordering information: Installation is required. Assembly videos available.

Spécifications
  • Désignation:
    2 shelf lab cart
  • Load Capacity:
    330 lbs.
Accessories

Informations sur le produit

  • Catalog #
    Availability
    Price
  • Extra Shelf for 4 shelf and 2 shelf trolleys
    Extra Shelf for 4 shelf and 2 shelf trolleys
    10129-048
    Log in pour la disponibilité
    Each - $449.10

Informations sur le produit

  • Catalog #
    Availability
    Price
  • End mesh panels (2)
    End mesh panels (2)
    10129-050
    Log in pour la disponibilité
    Each - $238.47