Treston Lab Carts | Avantor
Treston Lab Carts
4 shelf lab cart, 660 lbs.
Catalog # 10129-030
Treston Lab Carts
Catalog # 10129-030
Supplier:  
Treston Lab Carts
Treston Lab Carts
4 shelf lab cart, 660 lbs.
Discontinued
Product Details & Documents
TRTA lab carts are ideal for storing medical devices, instruments and supplies, and moving them to where they are needed.
  • Versatile and multi-purpose
  • Physically ergonomic for effortless cart movement
The height adjustment range of the shelves is 7.87 to 27.56".
 : Installation is required. Assembly videos available.
Spécifications
  • Désignation:
    4 shelf lab cart
  • Load Capacity:
    660 lbs.
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Extra Shelf for 4 shelf and 2 shelf trolleys
    Extra Shelf for 4 shelf and 2 shelf trolleys
    10129-048
    Log in pour la disponibilité
    Each - $449.10
Informations sur le produit
  • Catalog #
    Availability
    Price
  • End mesh panels (2)
    End mesh panels (2)
    10129-050
    Log in pour la disponibilité
    Each - $238.47