Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
TBS 10X Buffer Solution, pH 7.4, Quality Biological

TBS 10X Buffer Solution, pH 7.4, Quality Biological

TBS 10X pH 7.4, 1 L, 7.4 ±0.1
  10128-550
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Supplier #:  351-086-131
TBS 10X Buffer Solution, pH 7.4, Quality Biological
  10128-550
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
 :  
TBS 10X Buffer Solution, pH 7.4, Quality Biological
TBS 10X Buffer Solution, pH 7.4, Quality Biological

TBS 10X pH 7.4, 1 L, 7.4 ±0.1

 :  
 
$175.30
   
Each (1,000ml)
Minimum order quantity is 1
 

TBS 10X pH 7.4 is used as a washing buffer for cellular and molecular biology techniques.

  • DNase, RNase, and Protease tested

TBS 10X pH 7.4 will be useful as a washing buffer for electrodes, microtiter plates, and alkaline phophatase or peroxidase conjugates. TBS 10X pH 7.4 can also be used in detergent preparation and Western blot or ELISA applications.

Contenst: Tris base, Tris HCl, sodium chloride and Cell Culture Grade Water.

Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.

Caution: Research use only.

 

 

  • Désignation:
    TBS 10X pH 7.4
  • Taille:
    1 L
  • pH:
    7.4 ±0.1
  • Cat. No.:
    10128-550
  • Test description:
    DNAse, RNAse and protease tested
  • Shelf life:
    36 months
  • Sterility:
    Sterile filtered
  • Shipping temperature:
    Ambient

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.