Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Sodium Hydroxide

Sodium Hydroxide

Sodium Hydroxide, Laboratory, 500 g
Catalog # 470302-552
Supplier:   Ward's Science
Supplier #:  SS0550-500G
Sodium Hydroxide
Catalog # 470302-552
Supplier:  Ward's Science
CAS Number:  
Sodium Hydroxide
Sodium Hydroxide

Sodium Hydroxide, Laboratory, 500 g

CAS Number:  
Price
$29.05
Log in pour la disponibilité
Each (500g)
Minimum order quantity is 1
Product Details & Documents

CAS Number: 1310-73-2
Formula: NaOH
Density: 2.13 g/mL
Boiling and Freezing Point: 1390°C, 318°C
Solubility: Water and Alcohol
Synonyms: Caustic Soda
Shelf Life: 36 Months

Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.

Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Caution: This product is corrosive and toxic.

Spécifications

Spécifications

  • Désignation:
    Sodium Hydroxide
  • Taille:
    500 g
  • Chemical/Physical Specifications:
    Pellet
  • Grade:
    Laboratory

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.