Magnesium Hydroxide
Magnesium Hydroxide, Reagent, 500 g
Supplier: Ward's Science
Fournisseur #: MM0070-500GR
Jump to:
Product Details & Documents
CAS Number: 1309-42-8
Formula Weight: 58.32
Formula: Mg(OH)2
Solubility: Dilute Acids
Synonyms: Milk of Magnesia, Magnesium Hydrate
Shelf Life (months): 36
Storage: Green
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Spécifications
Spécifications
- Désignation:Magnesium Hydroxide
- Taille:500 g
- Chemical/Physical Specifications:Powder
- Grade:Reagent
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



