Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Fluid Thioglycollate, USP

Fluid Thioglycollate, USP

Fluid Thio Med, USP, 200 ml, Serum bottle, 3
Catalog # CA89407-581
Supplier #:  U207
Fluid Thioglycollate, USP
Catalog # CA89407-581
CAS Number:  
Fluid Thioglycollate, USP
Fluid Thioglycollate, USP

Fluid Thio Med, USP, 200 ml, Serum bottle, 3

CAS Number:  
Price
$446.00
Log in for availability
Pack of 10 (150ml)
Product Details & Documents

Fluid Thioglycollate Medium (FTM), is recommended for the use of of detecting microorganisms in sterile materials and USP sterility procedures.

Specifications

Specifications

  • Type de conditionnement:
    Serum bottle
  • Désignation:
    Fluid Thio Med, USP
  • Minimum Order Quantity:
    3
  • Volume:
    200 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.