Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Butterfield's Phosphate Buffer, Hardy Diagnostics

Butterfield's Phosphate Buffer, Hardy Diagnostics

pH 7.2, 12×100 ml, PP bottle
Catalog # CA89407-440
Supplier #:  U290
Butterfield's Phosphate Buffer, Hardy Diagnostics
Catalog # CA89407-440
CAS Number:  
Butterfield's Phosphate Buffer, Hardy Diagnostics
Butterfield's Phosphate Buffer, Hardy Diagnostics

pH 7.2, 12×100 ml, PP bottle

CAS Number:  
Price
$250.00
Log in pour la disponibilité
Pack of 12
Minimum order quantity is 1
Product Details & Documents

For the preparation of dilutions for plate count and other laboratory tests, pH 7.2.

Spécifications

Spécifications

  • Type de conditionnement:
    PP bottle
  • Désignation:
    pH 7.2
  • Taille:
    12×100 ml
  • Cat. No.:
    CA89407-440
  • Storage temperature:
    –2-30°C

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.