Butterfield's Phosphate Buffer, Hardy Diagnostics
pH 7.2, 20×9 ml, Glass tube, 20×125 mm
Supplier: HARDY DIAGNOSTICS CA
Supplier #: K209
Butterfield's Phosphate Buffer, Hardy Diagnostics
Catalog # CA89407-416
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
For the preparation of dilutions for plate count and other laboratory tests, pH 7.2.
Specifications
Specifications
- Type de conditionnement:Glass tube, 20×125 mm
- Désignation:pH 7.2
- Taille:20×9 ml
- Cat. No.:CA89407-416
- Storage temperature:–2-30°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



