Butterfield's Phosphate Buffer, Hardy Diagnostics
pH 7.2, 10×500 ml, PC bottle
Supplier: HARDY DIAGNOSTICS CA
Fournisseur #: U190
Butterfield's Phosphate Buffer, Hardy Diagnostics
Catalog # CA89407-414
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
For the preparation of dilutions for plate count and other laboratory tests, pH 7.2.
Spécifications
Spécifications
- Type de conditionnement:PC bottle
- Désignation:pH 7.2
- Taille:10×500 ml
- Cat. No.:CA89407-414
- Storage temperature:–2-30°C



