Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Butterfield's Phosphate Buffer, Hardy Diagnostics | Avantor

Butterfield's Phosphate Buffer, Hardy Diagnostics

pH 7.2, 20×9.9 ml, Glass tube, 16×125 mm
Catalog # CA89407-410
Fournisseur #:  K208
Butterfield's Phosphate Buffer, Hardy Diagnostics
Catalog # CA89407-410
CAS Number:  
Butterfield's Phosphate Buffer, Hardy Diagnostics
Butterfield's Phosphate Buffer, Hardy Diagnostics

pH 7.2, 20×9.9 ml, Glass tube, 16×125 mm

CAS Number:  
Price
$167.00
Log in pour la disponibilité
Pack of 20
La quantité minimale de commande est de 1
Product Details & Documents

For the preparation of dilutions for plate count and other laboratory tests, pH 7.2.

Spécifications

Spécifications

  • Type de conditionnement:
    Glass tube, 16×125 mm
  • Désignation:
    pH 7.2
  • Taille:
    20×9.9 ml
  • Cat. No.:
    CA89407-410
  • Storage temperature:
    –2-30°C

Spécifications