Butterfield's Phosphate Buffer, Hardy Diagnostics
pH 7.2, 20×9 ml, Glass tube, 16×125 mm
Supplier: HARDY DIAGNOSTICS CA
: K109
Butterfield's Phosphate Buffer, Hardy Diagnostics
Catalog # CA89407-408
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
For the preparation of dilutions for plate count and other laboratory tests, pH 7.2.
Spécifications
Spécifications
- Type de conditionnement:Glass tube, 16×125 mm
- Désignation:pH 7.2
- Taille:20×9 ml
- Cat. No.:CA89407-408
- Storage temperature:–2-30°C



