BCSA Base, CRITERION™, Hardy Diagnostics
10 kg, Bucket
  CA89406-818
 :  C8883
BCSA Base, CRITERION™, Hardy Diagnostics
  CA89406-818
 :  
BCSA Base, CRITERION™, Hardy Diagnostics
BCSA Base, CRITERION™, Hardy Diagnostics
10 kg, Bucket
 :  
$3,140.00
Each
CRITERION™ BCSA Base, used for the selective isolation of Burkholderia (Pseudomonas) cepacia
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Antibiotics Polymixin B, Gentamicin, and Vancomycin required. See product label for details. Dehydratedculture media with engineered container.
 : BSE Free
  • Type de conditionnement:
    Bucket
  • Taille:
    10 kg
  • Cat. No.:
    CA89406-818
  • Storage temperature:
    –2-30°C