BCSA Base, CRITERION™, Hardy Diagnostics
10 kg, Bucket
CA89406-818
: C8883
CRITERION™ BCSA Base, used for the selective isolation of Burkholderia (Pseudomonas) cepacia
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Antibiotics Polymixin B, Gentamicin, and Vancomycin required. See product label for details. Dehydratedculture media with engineered container.
: BSE Free
- Type de conditionnement:Bucket
- Taille:10 kg
- Cat. No.:CA89406-818
- Storage temperature:–2-30°C