BCSA Base, CRITERION™, Hardy Diagnostics
2 kg, Bucket
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C8882
BCSA Base, CRITERION™, Hardy Diagnostics
Catalog # CA89406-816
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ BCSA Base, used for the selective isolation of Burkholderia (Pseudomonas) cepacia
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Antibiotics Polymixin B, Gentamicin, and Vancomycin required. See product label for details. Dehydratedculture media with engineered container.
Certifications: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Bucket
- Taille:2 kg
- Cat. No.:CA89406-816
- Storage temperature:–2-30°C
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



