Modified EC Medium, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
  CA89406-680
 :  C8091
Modified EC Medium, CRITERION™, Hardy Diagnostics
  CA89406-680
 :  
Modified EC Medium, CRITERION™, Hardy Diagnostics
Modified EC Medium, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
 :  
$484.00
Each
CRITERION™ Modified EC Medium is for the detection of Escherichia coli 0157:H
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
 : BSE Free
  • Type de conditionnement:
    Bottle, wide mouth
  • Taille:
    500 g
  • Cat. No.:
    CA89406-680
  • Storage temperature:
    –2-30°C