CRITERION™ Tryptose, Hardy Diagnostics
500 g, Bottle, wide mouth
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C8071
CRITERION™ Tryptose, Hardy Diagnostics
Catalog # CA89406-668
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Tryptose is primarily used for preparing microbiological culture media and when non-animal source nutrients are needed in the cultivation of fastidious and non-fastidious organsims
- Non-mammalian
- Wide mouth opening
- Reduces dust and allows easy scooping
- Convenient handgrip
- 'No slip' finger holds
- Stackable containers
- High solubility; dissolves quickly
- Custom and bulk products available
Dehydrated culture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
Certifications: TSE/BSE Free
Specifications
Specifications
- Type de conditionnement:Bottle, wide mouth
- Taille:500 g
- Cat. No.:CA89406-668
- Storage temperature:–2-30°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



