Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

CRITERION™ Tryptose, Hardy Diagnostics

122.4 g, 2 L, Zip-pouch to make 2 L, Mylar®
Catalog # CA89406-666
CRITERION™ Tryptose, Hardy Diagnostics
Catalog # CA89406-666
Supplier:  
CAS Number:  
CRITERION™ Tryptose, Hardy Diagnostics
CRITERION™ Tryptose, Hardy Diagnostics

122.4 g, 2 L, Zip-pouch to make 2 L, Mylar®

CAS Number:  
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
TRYPTOSE AGAR 1X500G 500GM Catalog #CA95022-612

 

Product Details & Documents

CRITERION™ Tryptose is primarily used for preparing microbiological culture media and when non-animal source nutrients are needed in the cultivation of fastidious and non-fastidious organsims

  • Non-mammalian
  • Wide mouth opening
  • Reduces dust and allows easy scooping
  • Convenient handgrip
  • 'No slip' finger holds
  • Stackable containers
  • High solubility; dissolves quickly
  • Custom and bulk products available

Dehydrated culture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.

 : TSE/BSE Free

Spécifications

Spécifications

  • Type de conditionnement:
    Zip-pouch to make 2 L, Mylar®
  • Taille:
    122.4 g
  • Final Volume:
    2 L
  • Cat. No.:
    CA89406-666
  • Storage temperature:
    –2-30°C

Spécifications