CRITERION™ A-1 Medium, Hardy Diagnostics
2 kg, Bucket
: C7572
CRITERION™ A-1 Medium is recommended for the detection of fecal coliforms in foods, treated wastewater, and seawater.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
: BSE Free
- Type de conditionnement:Bucket
- Taille:2 kg
- Cat. No.:CA89406-408
- Storage temperature:–2-30°C