CRITERION™ A-1 Medium, Hardy Diagnostics
2 kg, Bucket
  CA89406-408
 :  C7572
CRITERION™ A-1 Medium, Hardy Diagnostics
  CA89406-408
 :  
CRITERION™ A-1 Medium, Hardy Diagnostics
CRITERION™ A-1 Medium, Hardy Diagnostics
2 kg, Bucket
 :  
$1,370.00
Each
CRITERION™ A-1 Medium is recommended for the detection of fecal coliforms in foods, treated wastewater, and seawater.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
 : BSE Free
  • Type de conditionnement:
    Bucket
  • Taille:
    2 kg
  • Cat. No.:
    CA89406-408
  • Storage temperature:
    –2-30°C