CRITERION™ A-1 Medium, Hardy Diagnostics
500 g, Bottle, wide mouth
Supplier: HARDY DIAGNOSTICS CA
: C7571
CRITERION™ A-1 Medium, Hardy Diagnostics
Catalog # CA89406-406
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ A-1 Medium is recommended for the detection of fecal coliforms in foods, treated wastewater, and seawater.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Certifications: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Bottle, wide mouth
- Taille:500 g
- Cat. No.:CA89406-406
- Storage temperature:–2-30°C
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



