Thioglycollate Medium without Indicator, CRITERION™, Hardy Diagnostics
2 kg, Bucket
Catalog # CA89406-138
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C7082
Thioglycollate Medium without Indicator, CRITERION™, Hardy Diagnostics
Catalog # CA89406-138
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Thioglycollate Medium without Indicator is recommended for the cultivation of aerobic, microaerophilic, and anaerobic microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
Specifications
- Type de conditionnement:Bucket
- Taille:2 kg
- Cat. No.:CA89406-138
- Storage temperature:–2-30°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



