Thioglycollate Medium without Indicator, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
Supplier: HARDY DIAGNOSTICS CA
Fournisseur #: C7081
Thioglycollate Medium without Indicator, CRITERION™, Hardy Diagnostics
Catalog # CA89406-136
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Thioglycollate Medium without Indicator is recommended for the cultivation of aerobic, microaerophilic, and anaerobic microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Bottle, wide mouth
- Taille:500 g
- Cat. No.:CA89406-136
- Storage temperature:–2-30°C
Spécifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



