Standard Methods Agar, CRITERION™, Hardy Diagnostics
10 kg, Bucket
Catalog # CA89406-068
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C6983
Standard Methods Agar, CRITERION™, Hardy Diagnostics
Catalog # CA89406-068
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Standard Methods Agar (Plate Count Agar) is recommended for use in determining the microbial content in dairy products, food, water and other material of sanitary importance
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Bucket
- Taille:10 kg
- Cat. No.:CA89406-068
- Storage temperature:–2-30°C



