SIM Medium, CRITERION™, Hardy Diagnostics
10 kg, Bucket
Catalog # CA89406-044
Supplier: HARDY DIAGNOSTICS CA
Fournisseur #: C6943
SIM Medium, CRITERION™, Hardy Diagnostics
Catalog # CA89406-044
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ SIM Medium is recommended for the differentiation of gram-negative enteric bacilli, such as Salmonella and Shigella, on the basis of sulfide production, indole formation, and motility
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Bucket
- Taille:10 kg
- Cat. No.:CA89406-044
- Storage temperature:–2-30°C
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



