SIM Medium, CRITERION™, Hardy Diagnostics
2 kg, Bucket
Catalog # CA89406-042
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C6942
SIM Medium, CRITERION™, Hardy Diagnostics
Catalog # CA89406-042
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ SIM Medium is recommended for the differentiation of gram-negative enteric bacilli, such as Salmonella and Shigella, on the basis of sulfide production, indole formation, and motility
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
Specifications
- Type de conditionnement:Bucket
- Taille:2 kg
- Cat. No.:CA89406-042
- Storage temperature:–2-30°C



