SIM Medium, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
: C6941
CRITERION™ SIM Medium is recommended for the differentiation of gram-negative enteric bacilli, such as Salmonella and Shigella, on the basis of sulfide production, indole formation, and motility
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
- Type de conditionnement:Bottle, wide mouth
- Taille:500 g
- Cat. No.:CA89406-040
- Storage temperature:–2-30°C