SIM Medium, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
  CA89406-040
 :  C6941
SIM Medium, CRITERION™, Hardy Diagnostics
  CA89406-040
 :  
SIM Medium, CRITERION™, Hardy Diagnostics
SIM Medium, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
 :  
$269.00
Each
CRITERION™ SIM Medium is recommended for the differentiation of gram-negative enteric bacilli, such as Salmonella and Shigella, on the basis of sulfide production, indole formation, and motility
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
  • Type de conditionnement:
    Bottle, wide mouth
  • Taille:
    500 g
  • Cat. No.:
    CA89406-040
  • Storage temperature:
    –2-30°C