Selective Strep Agar Base, CRITERION™, Hardy Diagnostics
10 kg, Bucket
CA89406-020
: C6893
CRITERION™ Selective Strep Agar Base with added sheep blood, this medium is recommended for the selective isolation and differentiation of pathogenic streptococci
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
- Type de conditionnement:Bucket
- Taille:10 kg
- Cat. No.:CA89406-020
- Storage temperature:–2-30°C