Selective Strep Agar Base, CRITERION™, Hardy Diagnostics
10 kg, Bucket
  CA89406-020
 :  C6893
Selective Strep Agar Base, CRITERION™, Hardy Diagnostics
  CA89406-020
 :  
Selective Strep Agar Base, CRITERION™, Hardy Diagnostics
Selective Strep Agar Base, CRITERION™, Hardy Diagnostics
10 kg, Bucket
 :  
$5,650.00
Each
CRITERION™ Selective Strep Agar Base with added sheep blood, this medium is recommended for the selective isolation and differentiation of pathogenic streptococci
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
 : BSE Free
  • Type de conditionnement:
    Bucket
  • Taille:
    10 kg
  • Cat. No.:
    CA89406-020
  • Storage temperature:
    –2-30°C