Selective Strep Agar Base, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
Supplier: HARDY DIAGNOSTICS CA
Fournisseur #: C6891
Selective Strep Agar Base, CRITERION™, Hardy Diagnostics
Catalog # CA89406-016
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Selective Strep Agar Base with added sheep blood, this medium is recommended for the selective isolation and differentiation of pathogenic streptococci
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Bottle, wide mouth
- Taille:500 g
- Cat. No.:CA89406-016
- Storage temperature:–2-30°C
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



