Nutrient Broth, CRITERION™, Hardy Diagnostics
122.4 g, Zip-pouch to make 2 L, Mylar®
Supplier: HARDY DIAGNOSTICS CA
: C6470
Nutrient Broth, CRITERION™, Hardy Diagnostics
Catalog # CA89405-792
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ Nutrient Broth is a general purpose growth media recommended for use in the cultivation of nonfastidious microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Zip-pouch to make 2 L, Mylar®
- Taille:122.4 g
- Cat. No.:CA89405-792
- Storage temperature:–2-30°C



