MIO Medium, CRITERION™, Hardy Diagnostics
2 kg, Bucket
Catalog # CA89405-730
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C6332
MIO Medium, CRITERION™, Hardy Diagnostics
Catalog # CA89405-730
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ MIO Medium is recommended for use in testing motility, indole production, and ornithine decarboxylase activity of enteric bacilli
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
Specifications
- Type de conditionnement:Bucket
- Taille:2 kg
- Cat. No.:CA89405-730
- Storage temperature:–2-30°C



