MIO Medium, CRITERION™, Hardy Diagnostics
500 g, Bottle, wide mouth
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C6331
MIO Medium, CRITERION™, Hardy Diagnostics
Catalog # CA89405-728
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ MIO Medium is recommended for use in testing motility, indole production, and ornithine decarboxylase activity of enteric bacilli
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
Specifications
- Type de conditionnement:Bottle, wide mouth
- Taille:500 g
- Cat. No.:CA89405-728
- Storage temperature:–2-30°C
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



