MacConkey Broth, CRITERION™, Hardy Diagnostics
122.4 g, 2 L, Zip-pouch to make 2 L, Mylar®
Supplier: HARDY DIAGNOSTICS CA
Supplier #: C6140
MacConkey Broth, CRITERION™, Hardy Diagnostics
Catalog # CA89405-636
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Jump to:
Product Details & Documents
CRITERION™ MacConkey Broth is recommended for cultivating gram-negative, lactose-fermenting bacilli from water and foods as a presumptive test for coliforms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
MacConkey Broth can also be used preenrichment of E. coli O157 for toxin testing. Dehydratedculture media with engineered container.
Certifications: BSE Free
Spécifications
Spécifications
- Type de conditionnement:Zip-pouch to make 2 L, Mylar®
- Taille:122.4 g
- Final Volume:2 L
- Cat. No.:CA89405-636
- Storage temperature:–2-30°C
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



